Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
CAS: 1330-20-7 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA447240010
Price (MYR)
380.00
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
380.00
Add to cart
Chemical Identifiers
CAS7772-99-8
IUPAC Nameλ²-tin(2+) dichloride
Molecular FormulaCl2Sn
InChI KeyAXZWODMDQAVCJE-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Iron (Fe)=<50 ppm
Appearance (Form)Crystalline powder or flakes
Lead (Pb)=<75 ppm
Titration Iodimetric97.5 to 102.5 %