Sodium bicarbonate, 99.5%, for biochemistry
Sodium bicarbonate, 99.5%, for biochemistry
Sodium bicarbonate, 99.5%, for biochemistry
Sodium bicarbonate, 99.5%, for biochemistry
Thermo Scientific Chemicals
Sodium bicarbonate, 99.5%, for biochemistry
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA447102500250 gPlastic Bottle
Catalog number FSA447102500
Price (MYR)
227.00
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
227.00
Add to cart
Specifications
Chemical Name or MaterialSodium bicarbonate
Melting Point270.0°C
CAS144-55-8
Quantity250 g
EINECS Number205-633-8
View more
Biological buffer
RUO – Research Use Only