Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA447080010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS14807-96-6
IUPAC Nametrimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate
Molecular FormulaH2Mg3O12Si4
InChI KeyKNXVOGGZOFOROK-UHFFFAOYSA-N
SMILES[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on ignition=<7.0 % (1 g, 1100°C)
Appearance (Color)White to off-white
Infrared spectrumConforms
Particle size=<75 µm
Loss on drying=<0.5 % (1 g, 105°C)