1-Chlorobutane, 99.5%, for analysis
1-Chlorobutane, 99.5%, for analysis
1-Chlorobutane, 99.5%, for analysis
1-Chlorobutane, 99.5%, for analysis
Thermo Scientific Chemicals
1-Chlorobutane, 99.5%, for analysis
CAS: 109-69-3 | C4H9Cl | 92.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4470200101 LGlass Bottle
Catalog number FSA447020010
Price (MYR)
688.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
688.00
Add to cart
Specifications
Chemical Name or Material1-Chlorobutane
CAS109-69-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o