tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Catalog number FSA447010010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
GradeAnalytical