Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number FSA446882500
Price (MYR)
1,431.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,431.00
Add to cart
Chemical Identifiers
CAS7631-99-4
IUPAC Namesodium nitrate
Molecular FormulaNNaO3
InChI KeyVWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES[Na+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPrills
Appearance (Color)White
Assay (unspecified)> 98.0%
CommentAs a bulk commodity material, may contain a small quantity of dark coloured prills from the manufacturing process
For HPLC
RUO – Research Use Only