Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number FSA446880500
Price (MYR)
413.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
413.00
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, sodium salt
CAS127-09-3
Melting Point324.0°C
EINECS Number204-823-8
Flash Point>250°C
View more
For HPLC
RUO – Research Use Only