Sodium chloride, 99.5%, for HPLC
Sodium chloride, 99.5%, for HPLC
Sodium chloride, 99.5%, for HPLC
Sodium chloride, 99.5%, for HPLC
Thermo Scientific Chemicals
Sodium chloride, 99.5%, for HPLC
CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA446212500250 gGlass bottle
Catalog number FSA446212500
Price (MYR)
972.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
972.00
Add to cart
Specifications
Boiling Point1461.0°C
Chemical Name or MaterialSodium chloride
Melting Point801.0°C
CAS7647-14-5
pH5.0 to 9.0