Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
CAS: 10378-23-1 | C10H12N2Na4O8·2H2O | 416.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA446085000
Price (MYR)
458.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
458.00
Add to cart
Chemical Identifiers
CAS59-67-6
IUPAC Namepyridine-3-carboxylic acid
Molecular FormulaC6H5NO2
InChI KeyPVNIIMVLHYAWGP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥98.5% (UK-sourced material)
CommentMaterial sourced in UK and US
Melting Point (clear melt)234.0-241.0°C
FormPowder
Appearance (Color)White to cream
View more
Chelation/Complexation reagent
RUO – Research Use Only