Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
CAS: 10378-23-1 | C10H12N2Na4O8·2H2O | 416.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA446081000
Price (MYR)
333.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
333.00
Add to cart
Chemical Identifiers
CAS12042-68-1
IUPAC Namecalcium dialumanoylolate
Molecular FormulaAl2CaO4
InChI KeyXFWJKVMFIVXPKK-UHFFFAOYSA-N
SMILES[Ca++].[O-][Al]=O.[O-][Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Chelation/Complexation reagent
RUO – Research Use Only