Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
CAS: 10378-23-1 | C10H12N2Na4O8·2H2O | 416.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA446080010
Price (MYR)
692.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
692.00
Add to cart
Chemical Identifiers
CAS112-61-8
IUPAC Namemethyl octadecanoate
Molecular FormulaC19H38O2
InChI KeyHPEUJPJOZXNMSJ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or Fused solid
Appearance (Color)White to cream
Assay (GC)>98.5%
Chelation/Complexation reagent
RUO – Research Use Only