Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Thermo Scientific Chemicals
Hydrogen chloride, 3M solution in CPME, AcroSeal™
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA445851000100 mLAcroSeal™ Glass bottle
Catalog number FSA445851000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS773-64-8
IUPAC Name2,4,6-trimethylbenzene-1-sulfonyl chloride
Molecular FormulaC9H11ClO2S
InChI KeyPVJZBZSCGJAWNG-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(C)=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)50.0-56.0?C