1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 872-50-4 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number FSA445790010
Price (MYR)
776.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
776.00
Add to cart
Specifications
Chemical Name or Material1-Methyl-2-pyrrolidinone
Melting Point-24.0°C
CAS872-50-4
Health Hazard 1GHS Signal Word: Danger
Density1.0300g/mL
View more
Suitable for battery materials development.
RUO – Research Use Only