n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA445720010
Price (MYR)
422.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
422.00
Add to cart
Chemical Identifiers
CAS11106-97-1
IUPAC Namechromium nickel
Molecular FormulaCrNi
InChI KeyVNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES[Cr].[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment0.2 mm diameter
CommentNi 80% min