Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA445670010
Price (MYR)
647.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
647.00
Add to cart
Chemical Identifiers
CAS2363-71-5
IUPAC Namehenicosanoic acid
Molecular FormulaC21H42O2
InChI KeyCKDDRHZIAZRDBW-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Glistening crystalline powder
GC>=98.5 %