Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Alfa Aesar
Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
FSA4452600505 g
Catalog number FSA445260050
Price (MYR)
1,405.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,405.00
Add to cart
Specifications
Chemical Name or MaterialNorfloxacin
TypeNorfloxacin
Physical FormCrystalline Powder
CAS70458-96-7
Melting Point218°C to 226°C