Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals
Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals
Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals
Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals
CAS: 5239-82-7 | C5H8O22 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA443780050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
FormFoil, 1.5mm (0.06in) thick.
Assay from Supplier's CofA≥99.998% (metals basis)