2-Amino-5-chloropyrimidine, 97%, Thermo Scientific™
2-Amino-5-chloropyrimidine, 97%, Thermo Scientific™
2-Amino-5-chloropyrimidine, 97%, Thermo Scientific™
2-Amino-5-chloropyrimidine, 97%, Thermo Scientific™
Thermo Scientific Chemicals
2-Amino-5-chloropyrimidine, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA443760050
Price (MYR)
686.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
686.00
Add to cart
Specifications
Chemical Name or Material2-Amino-5-chloropyrimidine
CAS5428-89-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Harmful if swallowed. Causes serious eye damage.
Health Hazard 3GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.