2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
CAS: 19767-45-4 | C2H5NaO3S2 | 164.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number FSA443150100
Price (MYR)
575.00
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
575.00
Add to cart
Specifications
Chemical Name or Material2-Mercaptoethanesulfonic acid sodium salt
CAS19767-45-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.