o-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
o-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
o-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
o-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
o-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
CAS: 95-47-6 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number FSA443021000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or Materialo-Xylene
Melting Point-25.0°C
CAS95-47-6
ColorWhite
Health Hazard 1Danger