Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number FSA443001000
Price (MYR)
272.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
272.00
Add to cart
Chemical Identifiers
CAS1873-88-7
IUPAC Name2,2,4,6,6-pentamethyl-3,5-dioxa-2,4,6-trisilaheptan-4-yl
Molecular FormulaC7H21O2Si3
InChI KeySWGZAKPJNWCPRY-UHFFFAOYSA-N
SMILESC[Si](O[Si](C)(C)C)O[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%