Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Catalog number FSA442980010
Price (MYR)
3,497.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
3,497.00
Add to cart
Chemical Identifiers
CAS119-80-2
IUPAC Name2-[(2-carboxyphenyl)disulfanyl]benzoic acid
Molecular FormulaC14H10O4S2
InChI KeyLBEMXJWGHIEXRA-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1SSC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %