4-Chlorophenethyl bromide, 95%
4-Chlorophenethyl bromide, 95%
4-Chlorophenethyl bromide, 95%
4-Chlorophenethyl bromide, 95%
Thermo Scientific Chemicals
4-Chlorophenethyl bromide, 95%
CAS: 6529-53-9 | C8H8BrCl | 219.51 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4401800505 gGlass bottle
Catalog number FSA440180050
Price (MYR)
842.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
842.00
Add to cart
Specifications
Chemical Name or Material4-Chlorophenethyl bromide
CAS6529-53-9
CAS Min %94.0
ColorColorless to Yellow
Health Hazard 1GHS Signal Word: Warning