2-Chlorophenethyl bromide, 95%, Thermo Scientific Chemicals
2-Chlorophenethyl bromide, 95%, Thermo Scientific Chemicals
2-Chlorophenethyl bromide, 95%, Thermo Scientific Chemicals
2-Chlorophenethyl bromide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorophenethyl bromide, 95%, Thermo Scientific Chemicals
CAS: 16793-91-2 | C8H8BrCl | 219.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA440140050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS63-68-3
IUPAC Name(2S)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular FormulaC5H11NO2S
InChI KeyFFEARJCKVFRZRR-BYPYZUCNSA-N
SMILESCSCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Loss on drying=<0.5 % (1 g, 105°C)
Specific optical rotation+22.4° to +24.7° (20°C, 589 nm) (c=2, 6 N HCl)
TLC=<0.5 % of any individual impurity