Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Thermo Scientific Chemicals
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA440061000100 mLAcroSeal™ Glass bottle
Catalog number FSA440061000
Price (MYR)
666.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
666.00
Add to cart
Chemical Identifiers
CAS141-53-7
IUPAC Namesodium formate
Molecular FormulaCHNaO2
InChI KeyHLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES[Na+].[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or pale cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1.0%
Phase transfer catalyst
For Manufacturing and Laboratory Use. Not for beverage use