3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
Alfa Aesar
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
CAS: 121-00-6 | C11H16O2 | 180.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number FSA439691000
Price (MYR)
210.61
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
210.61
Add to cart
Specifications
Chemical Name or Material3-tert-Butyl-4-hydroxyanisole
CAS121-00-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0