N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
Thermo Scientific Chemicals
N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
CAS: 95713-52-3 | C9H9FN4O5 | 272.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA439341000100 mgGlass bottle
Catalog number FSA439341000
Price (MYR)
2,084.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,084.00
Add to cart
Chemical Identifiers
CAS1674-38-0
IUPAC Name1-phenyldodecan-1-one
Molecular FormulaC18H28O
InChI KeyDJNJZIFFCJTUDS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)40.0-49.0?C
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥96.0%
Chiral derivatization reagent
RUO – Research Use Only