Isoprenaline hydrochloride, 98%, Thermo Scientific Chemicals
Isoprenaline hydrochloride, 98%, Thermo Scientific Chemicals
Isoprenaline hydrochloride, 98%, Thermo Scientific Chemicals
Isoprenaline hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isoprenaline hydrochloride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number FSA437210050
Price (MYR)
372.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
372.00
Add to cart
Specifications
Chemical Name or MaterialIsoprenaline hydrochloride
CAS51-30-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
View more