Gold, 99.99%, (trace metal basis), shot, Thermo Scientific Chemicals
Gold, 99.99%, (trace metal basis), shot, Thermo Scientific Chemicals
Gold, 99.99%, (trace metal basis), shot, Thermo Scientific Chemicals
Gold, 99.99%, (trace metal basis), shot, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold, 99.99%, (trace metal basis), shot, Thermo Scientific Chemicals
CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA437140050
Price (MYR)
10,721.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
10,721.00
Add to cart
Chemical Identifiers
CAS4965-36-0
IUPAC Name7-bromoquinoline
Molecular FormulaC9H6BrN
InChI KeyXYBSZCUHOLWQQU-UHFFFAOYSA-N
SMILESBrC1=CC=C2C=CC=NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
for ultra trace analysis
RUO – Research Use Only