Iron, 99.99%, (trace metal basis), granules, Thermo Scientific Chemicals
Iron, 99.99%, (trace metal basis), granules, Thermo Scientific Chemicals
Iron, 99.99%, (trace metal basis), granules, Thermo Scientific Chemicals
Iron, 99.99%, (trace metal basis), granules, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iron, 99.99%, (trace metal basis), granules, Thermo Scientific Chemicals
CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA437110050
Price (MYR)
426.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
426.00
Add to cart
Chemical Identifiers
CAS134-62-3
IUPAC NameN,N-diethyl-3-methylbenzamide
Molecular FormulaC12H17NO
InChI KeyMMOXZBCLCQITDF-UHFFFAOYSA-N
SMILESCCN(CC)C(=O)C1=CC=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<100 APHA
GC>=97.5 %
Infrared spectrumConforms
Water=<0.3 %
Appearance (Form)Clear liquid
View more
For ultra trace analysis, Reduction reagent
RUO – Research Use Only