Chromium nitrate nonahydrate, 99.99%, (trace metal basis)
Chromium nitrate nonahydrate, 99.99%, (trace metal basis)
Chromium nitrate nonahydrate, 99.99%, (trace metal basis)
Chromium nitrate nonahydrate, 99.99%, (trace metal basis)
Thermo Scientific Chemicals
Chromium nitrate nonahydrate, 99.99%, (trace metal basis)
CAS: 7789-02-8 | CrN3O9·9H2O | 400.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4371000505 gGlass bottle
Catalog number FSA437100050
Price (MYR)
509.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
509.00
Add to cart
Specifications
Chemical Name or MaterialChromium nitrate nonahydrate
Melting Point60.0°C
CAS7789-02-8
ColorBlack to Violet
Health Hazard 1GHS Signal Word: Warning
View more
for ultra trace analysis
RUO – Research Use Only