Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
FSA43678050050 gPlastic bottle
Catalog number FSA436780500
Price (MYR)
1,255.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,255.00
Add to cart
Chemical Identifiers
CAS2172-12-5
IUPAC Nametris(propan-2-ol) yttrium
Molecular FormulaC9H24O3Y
InChI KeyNREVZTYRXVBFAQ-UHFFFAOYSA-N
SMILES[Y].CC(C)O.CC(C)O.CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 90.0%
For ultra trace analysis
RUO – Research Use Only