Naproxen sodium, 98%, Thermo Scientific Chemicals
Naproxen sodium, 98%, Thermo Scientific Chemicals
Naproxen sodium, 98%, Thermo Scientific Chemicals
Naproxen sodium, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naproxen sodium, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number FSA436710050
Price (MYR)
431.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
431.00
Add to cart
Specifications
Chemical Name or MaterialNaproxen sodium
CAS26159-34-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
May damage fertility or the unborn child.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clo