4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
Thermo Scientific Chemicals
4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
CAS: 931-40-8 | C4H6O4 | 118.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA43617025025 gGlass bottle
Catalog number FSA436170250
Price (MYR)
1,463.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,463.00
Add to cart
Chemical Identifiers
CAS1314-62-1
IUPAC Name[(dioxovanadio)oxy]dioxovanadium
Molecular FormulaO5V2
InChI KeyGNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILESO=[V](=O)O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.