3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
CAS: 61272-71-7 | C7H6BrN3 | 212.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA435760010
Price (MYR)
678.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
678.00
Add to cart
Specifications
Chemical Name or Material3-Amino-5-bromo-1H-indazole
CAS61272-71-7
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0