3-Amino-1H-indazole, 95%
3-Amino-1H-indazole, 95%
3-Amino-1H-indazole, 95%
3-Amino-1H-indazole, 95%
3-Amino-1H-indazole, 95%
CAS: 874-05-5 | C7H7N3 | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA435740050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS399-95-1
IUPAC Name4-amino-3-fluorophenol
Molecular FormulaC6H6FNO
InChI KeyMNPLTKHJEAFOCA-UHFFFAOYSA-N
SMILESNC1=CC=C(O)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)134.0-144.0?C
Appearance (Color)Cream to brown
Assay (GC)≥95.0%