n-Nonane, 99%, anhydrous, AcroSeal™
n-Nonane, 99%, anhydrous, AcroSeal™
n-Nonane, 99%, anhydrous, AcroSeal™
n-Nonane, 99%, anhydrous, AcroSeal™
n-Nonane, 99%, anhydrous, AcroSeal™
Thermo Scientific Chemicals
n-Nonane, 99%, anhydrous, AcroSeal™
CAS: 111-84-2 | C9H20 | 128.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA435631000100 mLAcroSeal™ Glass Bottle
Catalog number FSA435631000
Price (MYR)
502.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
502.00
Add to cart
Specifications
Chemical Name or Materialn-Nonane
CAS111-84-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0