4-(Trifluoromethyl)phenylacetylene, 97%
4-(Trifluoromethyl)phenylacetylene, 97%
4-(Trifluoromethyl)phenylacetylene, 97%
4-(Trifluoromethyl)phenylacetylene, 97%
Thermo Scientific Chemicals
4-(Trifluoromethyl)phenylacetylene, 97%
CAS: 705-31-7 | C9H5F3 | 170.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4353700505 gGlass bottle
Catalog number FSA435370050
Price (MYR)
1,781.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,781.00
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethyl)phenylacetylene
CAS705-31-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0