3-chloro-4-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals
3-chloro-4-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals
3-chloro-4-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals
3-chloro-4-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals
Alfa Aesar
3-chloro-4-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals
CAS: 4903-09-7 | C8H7ClO2 | 170.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA435310010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-chloro-4-methoxybenzaldehyde
CAS4903-09-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0