Ethyl 2-cyanoacrylate
Ethyl 2-cyanoacrylate
Ethyl 2-cyanoacrylate
Ethyl 2-cyanoacrylate
Thermo Scientific Chemicals
Ethyl 2-cyanoacrylate
CAS: 7085-85-0 | C6H7NO2 | 125.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4344500505 gGlass bottle
Catalog number FSA434450050
Price (MYR)
1,083.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,083.00
Add to cart
Specifications
Chemical Name or MaterialEthyl 2-cyanoacrylate
CAS7085-85-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for