Trifluoroacetic acid, 99+%, for spectroscopy
Trifluoroacetic acid, 99+%, for spectroscopy
Trifluoroacetic acid, 99+%, for spectroscopy
Trifluoroacetic acid, 99+%, for spectroscopy
Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, for spectroscopy
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA434161000100 mLGlass bottle
Catalog number FSA434161000
Price (MYR)
1,348.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,348.00
Add to cart
Chemical Identifiers
CAS7790-80-9
IUPAC Namecadmium(2+) diiodide
Molecular FormulaCdI2
InChI KeyOKIIEJOIXGHUKX-UHFFFAOYSA-L
SMILES[Cd++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
For UV/VIS spectroscopy
RUO – Research Use Only