N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
Thermo Scientific Chemicals
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
CAS: 110-18-9 | C6H16N2 | 116.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA433831000100 mLAcroSeal™ Glass Bottle
Catalog number FSA433831000
Price (MYR)
573.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
573.00
Add to cart
Chemical Identifiers
CAS25085-99-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to pale yellow
FormLiquid or viscous liquid
Refractive Index1.500-1.640 at 20?C