4-Hydroxybenzenesulfonamide, 97%, Thermo Scientific Chemicals
4-Hydroxybenzenesulfonamide, 97%, Thermo Scientific Chemicals
4-Hydroxybenzenesulfonamide, 97%, Thermo Scientific Chemicals
4-Hydroxybenzenesulfonamide, 97%, Thermo Scientific Chemicals
Alfa Aesar
4-Hydroxybenzenesulfonamide, 97%, Thermo Scientific Chemicals
CAS: 1576-43-8 | C6H7NO3S | 1730.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA433300050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2075-45-8
IUPAC Name4-bromo-1H-pyrazole
Molecular FormulaC3H3BrN2
InChI KeyWVGCPEDBFHEHEZ-UHFFFAOYSA-N
SMILESBrC1=CNN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point92°C to 99°C
HPLC>=97.5 %
Appearance (Color)White to cream
Appearance (Form)Crystalline powder