Pentyltrichlorosilane, Thermo Scientific™
Pentyltrichlorosilane, Thermo Scientific™
Pentyltrichlorosilane, Thermo Scientific™
Pentyltrichlorosilane, Thermo Scientific™
Alfa Aesar
Pentyltrichlorosilane, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
FSA43310050050 gAcroSeal™ Glass Bottle
Catalog number FSA433100500
Price (MYR)
280.00
Add to cart
Quantity:
50 g
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
280.00
Add to cart
Specifications
Chemical Name or MaterialPentyltrichlorosilane
CAS107-72-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.