Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change view
Quantity:
100 mL
500 mL
Packaging:
View all
Glass bottle
Catalog number FSA432291000
Price (MYR)
2,200.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,200.00
Add to cart
Specifications
Absorbance0.03 max. at 300 nm, 0.03 max. at 320 nm, 0.04 max. at 290 nm, 0.05 max. at 280 nm, 0.1 max. at 270nm, 1 max. at 260 nm
Chemical Name or MaterialTrifluoroacetic acid
Name NoteFor HPLC
CAS76-05-1
Melting Point-15°C