Dodecyldimethylchlorosilane, 97%, Thermo Scientific Chemicals
Dodecyldimethylchlorosilane, 97%, Thermo Scientific Chemicals
Dodecyldimethylchlorosilane, 97%, Thermo Scientific Chemicals
Dodecyldimethylchlorosilane, 97%, Thermo Scientific Chemicals
Alfa Aesar
Dodecyldimethylchlorosilane, 97%, Thermo Scientific Chemicals
CAS: 66604-31-7 | C14H31ClSi | 262.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA432230250
Price (MYR)
310.90
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
310.90
Add to cart
Chemical Identifiers
CAS16419-60-6
IUPAC Name(2-methylphenyl)boronic acid
Molecular FormulaC7H9BO2
InChI KeyNSJVYHOPHZMZPN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5%
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
Appearance (Color)White to cream
CommentMay contain varying amounts of anhydride