3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
Thermo Scientific Chemicals
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
CAS: 24493-21-8 | C6H9D4NaO2Si | 172.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4321200101 gGlass bottle
Catalog number FSA432120010
Price (MYR)
1,230.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,230.00
Add to cart
Chemical Identifiers
CAS10326-27-9
IUPAC Namebarium(2+) dihydrate dichloride
Molecular FormulaBaCl2H4O2
InChI KeyPWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Solution Test(10% in water) : Clear & colorless (non-U.S. specification)
FormCrystalline powder
Appearance (Color)White to pale grey
Assay (Titration ex Chloride)≥99.0 to ≤101.0% (non-U.S. specification)
Assay from Supplier's CofA≥99.0% (U.S. specification)
For NMR
RUO – Research Use Only