kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
CAS: 11114-20-8 | C24H36O25S2-2 | 788.647 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA431590250
Price (MYR)
1,391.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,391.00
Add to cart
Specifications
Chemical Name or Materialkappa-Carrageenan
CAS11114-20-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful if inhaled.
Health Hazard 3GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.