Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
CAS: 106-92-3 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA430951000
Price (MYR)
271.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
271.00
Add to cart
Chemical Identifiers
CAS106930-51-2
IUPAC Nametert-butyl N-(1-hydroxy-4-methylpentan-2-yl)carbamate
Molecular FormulaC11H23NO3
InChI KeyLQTMEOSBXTVYRM-UHFFFAOYNA-N
SMILESCC(C)CC(CO)NC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%