Ethyl formate, 98+%, Extra Dry, AcroSeal™
Ethyl formate, 98+%, Extra Dry, AcroSeal™
Ethyl formate, 98+%, Extra Dry, AcroSeal™
Ethyl formate, 98+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Ethyl formate, 98+%, Extra Dry, AcroSeal™
CAS: 109-94-4 | C3H6O2 | 74.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA429241000100 mLAcroSeal™ Glass bottle
Catalog number FSA429241000
Price (MYR)
407.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
407.00
Add to cart
Chemical Identifiers
CAS1466-76-8
IUPAC Name2,6-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyMBIZFBDREVRUHY-UHFFFAOYSA-N
SMILESCOC1=CC=CC(OC)=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point187°C to 190°C
Titration with NaOH>=98.5 %